5-Bromoindole, 99%
5-Bromoindole, 99%
5-Bromoindole, 99%
Thermo Scientific Chemicals

5-Bromoindole, 99%

CAS: 10075-50-0 | C8H6BrN | 196.05 g/mol
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 107061000
Price (JPY)
-
Request A Quote
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10075-50-0
IUPAC Name5-bromo-1H-indole
Molecular FormulaC8H6BrN
InChI KeyVXWVFZFZYXOBTA-UHFFFAOYSA-N
SMILESBrC1=CC=C2NC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point89°C to 92°C
GC>=98.5 %
Appearance (Color)White to light brown
Infrared spectrumConforms
Appearance (Form)Powder and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only