Interested in a bulk or custom version of this product? Request pricing.

EPOR Recombinant Human Protein, hIgG1-Fc Tag

Catalog number: 10707H02H25

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Erythropoietin receptor (EPOR), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met1-Pro 250) of human EPOR (NP_000112.1) fused to the Fc region of human IgG1 at the C-terminus.

Activity: Measured by its binding ability in a functional ELISA . Immobilized CSF2RB/CD131 at 10 µg/ml(100 µl/well) can bind biotinylated recombinant human EPOR with a linear range of 0.16-4 µg/ml.

Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Cell Cycle & Proliferation, Other Research, Signal Transduction
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: EPOR, MGC138358
Gene ID: 2057
Gene Symbol: EPOR
Molecular Weight: 51.0 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Family: Hormones & Receptors
Protein Form: Recombinant, Truncated
Protein Name: Erythropoietin Receptor
Protein Subtype: Cytokine Receptors
Protein Tag or Fusion: IgG1 Fc Tag
Purity: ≥ 85% by SDS-PAGE
Species: Human
Validated Application: Assay Standard
Shipping Condition: Room Temperature

Contents & storage

Store at -20 to -70 °C

Documents

Manuals & protocols