1-Bromononane, 97%
1-Bromononane, 97%
1-Bromononane, 97%
1-Bromononane, 97%
Thermo Scientific Chemicals

1-Bromononane, 97%

CAS: 693-58-3 | C9H19Br | 207.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 107201000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS693-58-3
IUPAC Name1-bromononane
Molecular FormulaC9H19Br
InChI KeyAYMUQTNXKPEMLM-UHFFFAOYSA-N
SMILESCCCCCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to golden
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.4530 to 1.4550 (20°C, 589 nm)

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only