9-Bromophenanthrene, 96%
9-Bromophenanthrene, 96%
9-Bromophenanthrene, 96%
Thermo Scientific Chemicals

9-Bromophenanthrene, 96%

CAS: 573-17-1 | C14H9Br | 257.13 g/mol
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 107240250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS573-17-1
IUPAC Name9-bromophenanthrene
Molecular FormulaC14H9Br
InChI KeyRSQXKVWKJVUZDG-UHFFFAOYSA-N
SMILESBrC1=CC2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point60°C to 65°C
GC>=95 %
Appearance (Color)Light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only