DL-alpha-Bromophenylacetic acid, 97%
DL-alpha-Bromophenylacetic acid, 97%
DL-alpha-Bromophenylacetic acid, 97%
Thermo Scientific Chemicals

DL-alpha-Bromophenylacetic acid, 97%

CAS: 4870-65-9 | C8H7BrO2 | 215.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 107270250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4870-65-9
IUPAC Name2-bromo-2-phenylacetic acid
Molecular FormulaC8H7BrO2
InChI KeyWAKFRZBXTKUFIW-UHFFFAOYNA-N
SMILESOC(=O)C(Br)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Appearance (Color)White to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only