4-Bromophenylboronic acid, 98%
4-Bromophenylboronic acid, 98%
4-Bromophenylboronic acid, 98%
Thermo Scientific Chemicals

4-Bromophenylboronic acid, 98%

CAS: 5467-74-3 | C6H6BBrO2 | 200.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 107280050
Price (USD)
72.65
Special offer
Online exclusive
Ends: 28-Dec-2026
85.10
Save 12.45
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
72.65
Special offer
Online exclusive
Ends: 28-Dec-2026
85.10
Save 12.45
Each
Chemical Identifiers
CAS5467-74-3
IUPAC Name(4-bromophenyl)boronic acid
Molecular FormulaC6H6BBrO2
InChI KeyQBLFZIBJXUQVRF-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Titration with NaOH97.5 to 110.0 %
Appearance (Color)White to light yellow to light beige
Infrared spectrumConforms
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only