1-Bromopropane, 99%
1-Bromopropane, 99%
1-Bromopropane, 99%
Thermo Scientific Chemicals

1-Bromopropane, 99%

CAS: 106-94-5 | C3H7Br | 122.993 g/mol
数量:
250 mL
500 mL
1 L
パッケージ:
Glass bottle
製品番号(カタログ番号) 107312500
価格(JPY)
-
数量:
250 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS106-94-5
IUPAC Name1-bromopropane
Molecular FormulaC3H7Br
InChI KeyCYNYIHKIEHGYOZ-UHFFFAOYSA-N
SMILESCCCBr
さらに表示
Appearance (Color)Clear colorless to light yellow
GC>=98.5 %
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4320 to 1.4360 (20°C, 589 nm)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only