1-Bromopropane, 99%
1-Bromopropane, 99%
1-Bromopropane, 99%
Thermo Scientific Chemicals

1-Bromopropane, 99%

CAS: 106-94-5 | C3H7Br | 122.993 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
1 L
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 107315000
Price (EUR)
66,10
Each
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
66,10
Each
Chemical Identifiers
CAS106-94-5
IUPAC Name1-bromopropane
Molecular FormulaC3H7Br
InChI KeyCYNYIHKIEHGYOZ-UHFFFAOYSA-N
SMILESCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4320 to 1.4360 (20°C, 589 nm)
AppearanceMay darken on storage
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only