2-Bromopyridine, 99%
2-Bromopyridine, 99%
2-Bromopyridine, 99%
Thermo Scientific Chemicals

2-Bromopyridine, 99%

CAS: 109-04-6 | C5H4BrN | 158 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 107400250
Price (USD)
52.65
Special offer
Online exclusive
Ends: 28-Dec-2026
62.10
Save 9.45
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
52.65
Special offer
Online exclusive
Ends: 28-Dec-2026
62.10
Save 9.45
Each
Chemical Identifiers
CAS109-04-6
IUPAC Name2-bromopyridine
Molecular FormulaC5H4BrN
InChI KeyIMRWILPUOVGIMU-UHFFFAOYSA-N
SMILESBrC1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to light brown
GC>=98.5 %
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.5700 to 1.5730 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only