1X CD CHO Medium, 10 L - FAQs
View additional product information for CD CHO Medium - FAQs (10743002, 10743001)
1 product FAQs found
Does 1X CD CHO Medium contain a surfactant? Do I need to add extra surfactant for a suspension CHO culture in a shake flask?
While the full formulation for 1X CD CHO Medium is proprietary information that cannot be disclosed, we can confirm that the medium contains a surfactant and that no additional surfactant would need to be added to the medium to support your suspension CHO cells in shake flask culture.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.