4-Bromothiophenol, 95%
4-Bromothiophenol, 95%
4-Bromothiophenol, 95%
Thermo Scientific Chemicals

4-Bromothiophenol, 95%

CAS: 106-53-6 | C6H5BrS | 189.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 107460250
Price (EUR)
85,40
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
85,40
Each
Chemical Identifiers
CAS106-53-6
IUPAC Name4-bromobenzene-1-thiol
Molecular FormulaC6H5BrS
InChI KeyFTBCOQFMQSTCQQ-UHFFFAOYSA-N
SMILESSC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to beige
Melting point71°C to 76°C
Appearance (Form)Crystalline powder or crystals
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only