4-Bromothiophenol, 95%
4-Bromothiophenol, 95%
4-Bromothiophenol, 95%
Thermo Scientific Chemicals

4-Bromothiophenol, 95%

CAS: 106-53-6 | C6H5BrS | 189.07 g/mol
数量:
5 g
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 107461000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS106-53-6
IUPAC Name4-bromobenzene-1-thiol
Molecular FormulaC6H5BrS
InChI KeyFTBCOQFMQSTCQQ-UHFFFAOYSA-N
SMILESSC1=CC=C(Br)C=C1
さらに表示
Infrared spectrumConforms
Appearance (Color)White to beige
Melting point71°C to 76°C
Appearance (Form)Crystalline powder or crystals
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only