5-Bromo-DL-tryptophan, 99%
5-Bromo-DL-tryptophan, 99%
5-Bromo-DL-tryptophan, 99%
Thermo Scientific Chemicals

5-Bromo-DL-tryptophan, 99%

CAS: 6548-09-0 | C11H11BrN2O2 | 283.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
5 g
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 107510050
Price (TWD)
-
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS6548-09-0
IUPAC Name2-amino-3-(5-bromo-1H-indol-3-yl)propanoic acid
Molecular FormulaC11H11BrN2O2
InChI KeyKZDNJQUJBMDHJW-UHFFFAOYSA-N
SMILESNC(CC1=CNC2=CC=C(Br)C=C12)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige to light grey
Titration with HClO498.5 to 101.5 %
Infrared spectrumConforms
Appearance (Form)Powder or crystalline powder
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only