Butadiene sulfone, 98%
Butadiene sulfone, 98%
Butadiene sulfone, 98%
Thermo Scientific Chemicals

Butadiene sulfone, 98%

CAS: 77-79-2 | C4H6O2S | 118.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
2.5 kg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 107611000
Price (USD)
29.65
Online Exclusive
33.30
Save 3.65 (11%)
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
29.65
Online Exclusive
33.30
Save 3.65 (11%)
Each
Chemical Identifiers
CAS77-79-2
IUPAC Name2,5-dihydro-1λ⁶-thiophene-1,1-dione
Molecular FormulaC4H6O2S
InChI KeyMBDNRNMVTZADMQ-UHFFFAOYSA-N
SMILESO=S1(=O)CC=CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Adhering powder or crystalline powder
Appearance (Form)or crystals and/or chunks
Infrared spectrumConforms
Melting point63°C to 67°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only