2,3-Butanediol, 98%, mixture of racemic and meso forms, techn.
2,3-Butanediol, 98%, mixture of racemic and meso forms, techn.
2,3-Butanediol, 98%, mixture of racemic and meso forms, techn.
Thermo Scientific Chemicals

2,3-Butanediol, 98%, mixture of racemic and meso forms, techn.

CAS: 513-85-9 | C4H10O2 | 90.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 107640025
Price (EUR)
494,00
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
494,00
Each
Specifications
Assay Percent Range97.5% min. (GC)
Beilstein01,479
CAS513-85-9
ChEBICHEBI:62064
Chemical Name or Material2, 3-Butanediol
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only