2,3-Butanediol, 98%, mixture of racemic and meso forms, techn.
2,3-Butanediol, 98%, mixture of racemic and meso forms, techn.
2,3-Butanediol, 98%, mixture of racemic and meso forms, techn.
Thermo Scientific Chemicals

2,3-Butanediol, 98%, mixture of racemic and meso forms, techn.

CAS: 513-85-9 | C4H10O2 | 90.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 107642500
Price (USD)
279.65
Online Exclusive
311.00
Save 31.35 (10%)
Each
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
279.65
Online Exclusive
311.00
Save 31.35 (10%)
Each
Chemical Identifiers
CAS513-85-9
IUPAC Name(2R,3S)-butane-2,3-diol
Molecular FormulaC4H10O2
InChI KeyOWBTYPJTUOEWEK-ZXZARUISSA-N
SMILESC[C@@H](O)[C@H](C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4330 to 1.4390 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only