2,3-Butanedione, 99%
2,3-Butanedione, 99%
2,3-Butanedione, 99%
Thermo Scientific Chemicals

2,3-Butanedione, 99%

CAS: 431-03-8 | C4H6O2 | 86.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 107650050
Price (USD)
58.80
Each
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
58.80
Each
Chemical Identifiers
CAS431-03-8
IUPAC Namebutane-2,3-dione
Molecular FormulaC4H6O2
InChI KeyQSJXEFYPDANLFS-UHFFFAOYSA-N
SMILESCC(=O)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Specific gravity(20°C) 0.98 to 0.99
Infrared spectrumConforms
Refractive index1.3910 to 1.3990 (20°C, 589 nm)
Appearance (Color)Clear yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only