sec-Butanol, 99%, Extra Pure
sec-Butanol, 99%, Extra Pure
sec-Butanol, 99%, Extra Pure
Thermo Scientific Chemicals

sec-Butanol, 99%, Extra Pure

CAS: 78-92-2 | C4H10O | 74.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass Bottle
Catalog number 107700025
Price (EUR)
43,65
Online exclusive
48,20
Save 4,55 (9%)
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
43,65
Online exclusive
48,20
Save 4,55 (9%)
Each
Chemical Identifiers
CAS78-92-2
IUPAC Namebutan-2-ol
Molecular FormulaC4H10O
InChI KeyBTANRVKWQNVYAZ-UHFFFAOYNA-N
SMILESCCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Refractive index1.3960 to 1.3980 (20°C, 589 nm)
Appearance (Form)Clear liquid
Infrared spectrumConforms
Water=<0.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only