tert-Butanol, 99.5%, extra pure
tert-Butanol, 99.5%, extra pure
tert-Butanol, 99.5%, extra pure
Thermo Scientific Chemicals

tert-Butanol, 99.5%, extra pure

CAS: 75-65-0 | C4H10O | 74.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Plastic Drum
Catalog number 107710100
Price (USD)
289.65
Online Exclusive
322.00
Save 32.35 (10%)
Each
Quantity:
10 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (USD)
289.65
Online Exclusive
322.00
Save 32.35 (10%)
Each
Chemical Identifiers
CAS75-65-0
IUPAC Name2-methylpropan-2-ol
Molecular FormulaC4H10O
InChI KeyDKGAVHZHDRPRBM-UHFFFAOYSA-N
SMILESCC(C)(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.03 % (K.F.)
Appearance (Color)Clear colorless
Refractive index1.3860 to 1.3880 (20°C, 589 nm)
Appearance (Form)Liquid after melting
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only