tert-Butyl acetate, 99%
tert-Butyl acetate, 99%
tert-Butyl acetate, 99%
Thermo Scientific Chemicals

tert-Butyl acetate, 99%

CAS: 540-88-5 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 107765000
Price (EUR)
80,65
Online exclusive
89,30
Save 8,65 (10%)
Each
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
80,65
Online exclusive
89,30
Save 8,65 (10%)
Each
Chemical Identifiers
CAS540-88-5
IUPAC Nametert-butyl acetate
Molecular FormulaC6H12O2
InChI KeyWMOVHXAZOJBABW-UHFFFAOYSA-N
SMILESCC(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.3850 to 1.3880 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only