tert-Butylamine, 99%
tert-Butylamine, 99%
tert-Butylamine, 99%
Thermo Scientific Chemicals

tert-Butylamine, 99%

CAS: 75-64-9 | C4H11N | 73.13 g/mol
数量:
5 mL
1 L
2.5 L
10 L
パッケージ:
すべて表示
Glass Bottle
製品番号(カタログ番号) 107820025
価格(JPY)
-
数量:
2.5 L
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS75-64-9
IUPAC Name2-methylpropan-2-amine
Molecular FormulaC4H11N
InChI KeyYBRBMKDOPFTVDT-UHFFFAOYSA-N
SMILESCC(C)(C)N
さらに表示
Appearance (Form)Clear liquid
Color scale=<25 APHA
Infrared spectrumConforms
GC>=98.5 %
Water=<0.10 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only