DEF KER GROWTH SUPP - FAQs
View additional product information for DEF KER GROWTH SUPP - FAQs (10784015)
4 product FAQs found
Are the Defined Keratinocyte SFM basal medium and supplement available separately?
The Defined Keratinocyte SFM basal medium and supplement are not available separately. The kit can be ordered with catalog #10744-019.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Are the Keratinocyte SFM basal medium and supplement kit available separately?
The basal medium is not available for purchase individually, and can only be ordered as part of the Keratinocyte SFM kit (catalog # 17005-042). The supplements are available separately: Bovine Pituitary Extract is catalog # 13028-014 and the EGF is catalog # PHG0311.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Which serum-free media can be used to culture mouse keratinocytes?
The Keratinocyte SFM without calcium chloride (Cat. No. 37010-022) is appropriate for the culture of mouse keratinocytes. The medium should be supplemented with calcium chloride at a final concentration of 0.02 mM.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Which serum-free media can be used to culture human keratinocytes?
There are a variety of options for serum-free culture of human keratinocytes. Keratinocyte SFM (Cat. No. 17005-042) is a widely used kit that includes the basal medium, bovine pituitary extract (BPE) and Epidermal Growth Factor (EGF). If you would like a serum-free, defined culture system, the options are the Defined Keratinocyte SFM kit (Cat. No. 10744-019) or Epilife medium (Cat. No. M-EPI-500-CA) supplemented with EDGS (Cat. No. S-012-5). If you would like an animal origin free defined system, Epilife medium (Cat. No. M-EPI-500-CA) with S7 supplement (Cat. No. S-017-5) is an excellent choice.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.