n-Butylbenzene, 99+%
n-Butylbenzene, 99+%
n-Butylbenzene, 99+%
Thermo Scientific Chemicals

n-Butylbenzene, 99+%

CAS: 104-51-8 | C10H14 | 134.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number 107850010
Price (USD)
468.65
Online exclusive
521.00
Save 52.35
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
468.65
Online exclusive
521.00
Save 52.35
Each
Chemical Identifiers
CAS104-51-8
IUPAC Namebutylbenzene
Molecular FormulaC10H14
InChI KeyOCKPCBLVNKHBMX-UHFFFAOYSA-N
SMILESCCCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.4880 to 1.4900 (20°C, 589 nm)
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only