Are the Defined Keratinocyte SFM basal medium and supplement available separately?
The Defined Keratinocyte SFM basal medium and supplement are not available separately. The kit can be ordered with catalog #10744-019.