Second Strand Buffer - FAQs
View additional product information for Second Strand Buffer - FAQs (10812014)
1 product FAQs found
I am going to be producing cDNA from RNA. I have purchased the SuperScript II RT and would like to get some of the 5X Second Strand Buffer. Do you have a catalog number for it?
Yes, 5X Second Strand Buffer is available as a stand-alone product. The catalog number is 10812-014.