n-Butyric Anhydride, 98%
n-Butyric Anhydride, 98%
n-Butyric Anhydride, 98%
Thermo Scientific Chemicals

n-Butyric Anhydride, 98%

CAS: 106-31-0 | C8H14O3 | 158.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Packaging:
View all
Glass Bottle
Catalog number 108125000
Price (USD)
43.65
Special offer
Online exclusive
Ends: 30-Jun-2026
51.10
Save 7.45 (15%)
Each
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
43.65
Special offer
Online exclusive
Ends: 30-Jun-2026
51.10
Save 7.45 (15%)
Each
Chemical Identifiers
CAS106-31-0
IUPAC Namebutanoyl butanoate
Molecular FormulaC8H14O3
InChI KeyYHASWHZGWUONAO-UHFFFAOYSA-N
SMILESCCCC(=O)OC(=O)CCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4120 to 1.4140 (20°C, 589 nm)
GC>=97.5 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only