gamma-Butyrolactone, 99+
Product Image
Thermo Scientific Chemicals

gamma-Butyrolactone, 99+

CAS: 96-48-0 | C4H6O2 | 86.09 g/mol
製品番号(カタログ番号) 108130010
価格(JPY)
-
見積もりを依頼する
数量:
1 L
一括またはカスタム形式をリクエストする
化学物質識別子
CAS96-48-0
IUPAC Nameoxolan-2-one
Molecular FormulaC4H6O2
InChI KeyYEJRWHAVMIAJKC-UHFFFAOYSA-N
SMILESO=C1CCCO1
さらに表示
Color scale=<40 APHA
Refractive index1.4350 to 1.4375 (20°C, 589 nm)
Infrared spectrumConforms
Water=<0.1 % (K.F.)
Appearance (Form)Clear liquid
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only