n-Butyronitrile, 99%
n-Butyronitrile, 99%
n-Butyronitrile, 99%
Thermo Scientific Chemicals

n-Butyronitrile, 99%

CAS: 109-74-0 | C4H7N | 69.11 g/mol
Quantity:
100 mL
500 mL
2.5 L
Catalog number 108140025
Price (JPY)
-
Quantity:
2.5 L
Request bulk or custom format
Chemical Identifiers
CAS109-74-0
IUPAC Namebutanenitrile
Molecular FormulaC4H7N
InChI KeyKVNRLNFWIYMESJ-UHFFFAOYSA-N
SMILESCCCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Infrared spectrumConforms
Refractive index1.3830 to 1.3850 (20°C, 589 nm)
Color scale=<25 APHA
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only