Interested in a bulk or custom version of this product? Request pricing.

APOE Recombinant Human Protein, N-TRX.His tag

Catalog number: 10817H30E25

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Apolipoprotein E (APOE), recombinant human protein is supplied as a frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human APOE (P02649) (Lys 19-His 317) fused to a Trx and a polyhistidine tag at the N-terminus.

Activity: The activity of this protein has not been tested.

Formulation: PBS, 40% Glycerol, 0.5 mM TCEP, pH 7.4.

Reconstitution: No reconstitution needed. Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Expression: E. coli
Form: Frozen
Gene Aliases: AD2, LDLCQ5, LPG, MGC1571
Gene ID: 348
Gene Symbol: APOE
Molecular Weight: 51.3 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Family: Other Proteins
Protein Form: Full Length, Recombinant
Protein Name: Apolipoprotein E
Protein Subtype: Other Proteins
Protein Tag or Fusion: His Tag, Thioredoxin
Purification: PAGE
Purity: ≥ 90% by SDS-PAGE
Research Category: Inflammation
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 5 vials of protein in suspension buffer. Store at -80 °C.

Documents

Manuals & protocols