D(+)-10-Camphorsulfonic acid, 99%
D(+)-10-Camphorsulfonic acid, 99%
D(+)-10-Camphorsulfonic acid, 99%
Thermo Scientific Chemicals

D(+)-10-Camphorsulfonic acid, 99%

CAS: 3144-16-9 | C10H15O4S | 231.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 108225000
Price (USD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS3144-16-9
IUPAC Name[(1R,4S)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonate
Molecular FormulaC10H15O4S
InChI KeyMIOPJNTWMNEORI-XVKPBYJWSA-M
SMILESCC1(C)[C@H]2CC[C@]1(CS([O-])(=O)=O)C(=O)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Melting point>=190°C
Loss on drying=<2 % (1 g, 105°C, 5 hrs)
Sulfate (SO4)=<500 ppm
Titration with NaOH98.5 to 101.5 % (On dried substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only