Le milieu KnockOut™ D-MEM est un milieu basal optimisé pour la croissance de cellules souches pluripotentes embryonnaires et induites nonAfficher plus
Have Questions?
Référence
Quantité
10829018
500 mL
Référence 10829018
Prix (EUR)
32,34
Each
Quantité:
500 mL
Customize this product
Prix (EUR)
32,34
Each
Le milieu KnockOut™ D-MEM est un milieu basal optimisé pour la croissance de cellules souches pluripotentes embryonnaires et induites non différenciées (1). L’osmolarité est optimisée pour se rapprocher de celle du tissu embryonnaire de la souris.
Ne contient pas de L-glutamine.
Usage exclusivement réservé à la recherche. Ne pas utiliser pour des procédures de diagnostic.
Spécifications
Type de celluleCellules souches (embryonnaires), cellules souches (iPS - souches pluripotentes induites)
Qualité de fabricationBPFa pour dispositifs médicaux, 21 CFR Part 820 et ISO 13485
Gamme de produitsKnockOut
Type de produitKnockOut DMEM
Quantité500 mL
Conditions d’expéditionTempérature ambiante
FormeLiquide
Avec additifsHaute teneur en glucose, Rouge de phénol
Sans additifSans glutamine
Unit SizeEach
Contenu et stockage
Stocker au réfrigérateur (2 – 8°C) à l’abri de la lumière.
Foire aux questions (FAQ)
Can KnockOut SR be used to culture mouse ESCs in feeder-free conditions?
Yes, mouse embryonic stem cells can be cultured using KnockOut SR under feeder-free conditions. Typically, cells are plated at a higher seeding density, in the presence of LIF, on a 0.1% gelatin layer in place of feeder cells.
How do I characterize human embryonic stem ( ES) cells?
Human ES cells are generally characterized by their typical morphology (they grow as tightly packed clusters of small cells with high ratio of nucleus to cytoplasm); surface marker expression; RT-PCR detection of stem cell-specific gene expression (such as Oct3/4, Sox2, and Nanog); alkaline phosphatase staining, and telomerase activity assay. The most commonly used ES specific surface markers include stage-specific embryonic antigens SSEA-3 and SSEA-4 for human ES cells. Other ES-specific surface antigens also include TRA-1-60 and TRA-1-81. (Science 282:1145 (1998).
How are human embryonic stem (ES) cells derived?
Human ES cells are derived from human blastocyst inner cell masses, isolated by immunosurgery with rabbit antiserum to BeWO cells (a human trophoblast cell line) (Science 282:1145 (1998)).
What are ES cells?
Embryonic stem (ES) cells are derived from the early mammalian embryo and are capable of unlimited, undifferentiated proliferation in vitro while maintaining their potential to differentiate into a wide of range of adult tissues including germ cells. The pluripotency of the ES cells is normally demonstrated in vitro by inducing ES cells to differentiate into embryoid bodies and checking lineage-specific markers for differentiated cells in three body layers (endo, meso, and ectoderm), or injecting them into immunodeficient mice and determining the cell types produced in the teratomas.