(Carbethoxymethylene)triphenylphosphorane, 98+%
(Carbethoxymethylene)triphenylphosphorane, 98+%
(Carbethoxymethylene)triphenylphosphorane, 98+%
Thermo Scientific Chemicals

(Carbethoxymethylene)triphenylphosphorane, 98+%

CAS: 1099-45-2 | C22H21O2P | 348.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 108291000
Price (USD)
262.65
Special offer
Online exclusive
Ends: 28-Dec-2026
309.00
Save 46.35
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
262.65
Special offer
Online exclusive
Ends: 28-Dec-2026
309.00
Save 46.35
Each
Chemical Identifiers
CAS1099-45-2
IUPAC Nameethyl 2-(triphenyl-λ⁵-phosphanylidene)acetate
Molecular FormulaC22H21O2P
InChI KeyIIHPVYJPDKJYOU-UHFFFAOYSA-N
SMILESCCOC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point124°C to 129°C
Appearance (Color)White to light yellow
Infrared spectrumConforms
HPLC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
RUO – Research Use Only