Thermo Scientific Chemicals

N-Carbobenzyloxyglycine, 98.5%, Thermo Scientific Chemicals

Catalog number: 108350250
25 g, Plastic Bottle, Each
Thermo Scientific Chemicals

N-Carbobenzyloxyglycine, 98.5%, Thermo Scientific Chemicals

Catalog number: 108350250
25 g, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 108350250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1138-80-3
IUPAC Name
2-{[(benzyloxy)carbonyl]amino}acetic acid
Molecular Formula
C10H11NO4
InChI Key
CJUMAFVKTCBCJK-UHFFFAOYSA-N
SMILES
OC(=O)CNC(=O)OCC1=CC=CC=C1
Melting point
117°C to 122°C
HPLC
>=98.0 %
Titration with NaOH
>=98.0 %
Appearance (Color)
White
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text