LB Broth (1X)
Gibco™

LB Broth (1X)

LB broth (Lennox formulation) is a ready-to-use liquid growth medium used for bacterial culture. Gibco LB Broth is filter-sterilized andRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
10855021500 mL
108550011 L
Catalog number 10855021
Price (USD)
40.65
Online exclusive
44.24
Save 3.59
Each
Quantity:
500 mL
Price (USD)
40.65
Online exclusive
44.24
Save 3.59
Each

LB broth (Lennox formulation) is a ready-to-use liquid growth medium used for bacterial culture. Gibco LB Broth is filter-sterilized and can be used for nearly all applications that require a liquid bacteria culturing step, such as protein expression, cloning, and expansion.

LB broth is one of the most referenced bacterial media for cultivating and maintaining E. coli strains. Our LB broth is based on the Lennox formulation, one of the most commonly used formulations for LB medium.

LB broth formulation
Formulation per liter: 10 g SELECT Peptone 140, 5 g SELECT Yeast Extract, 5 g sodium chloride.
• Tryptone provides nitrogen, vitamins, minerals, and amino acids essential for growth
• Yeast extract is a source of vitamins, particularly B-group vitamins
• Sodium chloride supplies essential electrolytes for transport and osmotic balance

Primary applications
• Growth and maintenance of E. coli strains for plasmid purification and/or protein expression
• Resuspension of T. pyriformis cell fractions

Gibco media bottles
Each media bottle features an angled opening for simplified pipetting. The 1-L Gibco media bottle uses 39% less source material (33% less for the 500-mL bottle) than comparable media bottles on the market. The cap and bottle are made from high-density polyethylene (HDPE) and polyethylene terephthalate (PETE) and can both be recycled.

Find the media format that you need
We offer other ready-to-use media formulations, such as Terrific Broth or S.O.C Recovery Medium.
Explore other media formats for bacterial growth.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatBottle
Media TypeLB Broth (Lennox)
Preparation MethodReady-to-Use
Quantity500 mL
Shipping ConditionRoom Temperature
Product TypeLiquid Medium
SpeciesEscherichia coli
SterilitySterile
Target Organism ClassColiforms
Unit SizeEach
Contents & Storage
• 500 mL LB Broth (1X)

Store at room temperature.

Frequently asked questions (FAQs)

What additional materials do I need for my transformation?

You will need bacterial growth media, such as LB, the bacterial selection antibiotic, and induction reagent (IPTG, X-gal). We offer imMedia growth medium which is a pre-mixed, pre-sterilized medium containing selection antibiotics for growth of E. coli strains. It can be prepared with or without IPTG and X-gal.

What bacterial growth media should I use?

Check out our bacterial growth media chart (https://www.thermofisher.com/us/en/home/life-science/cell-culture/microbiological-culture/bacterial-growth-media.html).

What is the optimal pH of low salt LB for LB + blasticidin plates?

We recommend a pH of 7 or less and half the normal amount of NaCl in your LB media or plates.

See the following paper for details on optimal conditions: Yamaguchi et al (1965) Inhibition of Protein Synthesis by Blasticidin S. Journal of Biochemistry (Tokyo) Volume 57: pp 667-677.