3-chlorobenzaldehyde, 99%
3-chlorobenzaldehyde, 99%
3-chlorobenzaldehyde, 99%
Thermo Scientific Chemicals

3-chlorobenzaldehyde, 99%

CAS: 587-04-2 | C7H5ClO | 140.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 108610250
Price (EUR)
35,65
Online exclusive
39,10
Save 3,45
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
35,65
Online exclusive
39,10
Save 3,45
Each
Chemical Identifiers
CAS587-04-2
IUPAC Name3-chlorobenzaldehyde
Molecular FormulaC7H5ClO
InChI KeySRWILAKSARHZPR-UHFFFAOYSA-N
SMILESClC1=CC=CC(C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Refractive index1.5630 to 1.5650 (20°C, 589 nm)
GC>=98.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only