3-Chlorobenzoic acid, 99+%
3-Chlorobenzoic acid, 99+%
3-Chlorobenzoic acid, 99+%
Thermo Scientific Chemicals

3-Chlorobenzoic acid, 99+%

CAS: 535-80-8 | C7H5ClO2 | 156.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number 108660250
Price (USD)
30.65
Online exclusive
34.20
Save 3.55
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
30.65
Online exclusive
34.20
Save 3.55
Each
Chemical Identifiers
CAS535-80-8
IUPAC Name3-chlorobenzoic acid
Molecular FormulaC7H5ClO2
InChI KeyLULAYUGMBFYYEX-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
HPLC>=99.0 %
Appearance (Color)White to beige
Infrared spectrumConforms
Titration with NaOH99.0 to 100.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only