Interested in a bulk or custom version of this product? Request pricing.

LAIR2 Recombinant Human Protein

Catalog number: 10868HNAH25

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Leukocyte-Associated Immunoglobulin-Like Receptor 2 (LAIR2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications

This recombinant protein was expressed from a DNA sequence encoding the human LAIR2 (NP_002279.2) (Met 1-Pro 152).

Activity: Measured by the ability of the immobilized protein to support the adhesion of HT-29 human colon adenocarcinoma cells. When 5 x 104 cells/well are added to recombiannt human LAIR2 coated plates (50 µg/ml with 100 µl/well), approximately 30% will adhere after 30 minutes at 37°C. Optimal concentration depends on cell type as well as the application or research objectives.

Formulation: Lyophilized in 50 mM Tris, 0.7M NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: CD Proteins
Research Category: Immunology
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: LAIR2, XXbac-BCX535A19.6, CD306, MGC71634
Gene Symbol: LAIR2
Molecular Weight: 14.1 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Name: leukocyte-associated immunoglobulin-like receptor 2
Protein Subtype: Other Proteins
Protein Tag or Fusion: Untagged
Purity: ≥ 90% by SDS-PAGE
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 5 vial of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols