Detect rhabdovirus contamination in Sf9 cultures. Learn more »
Sf-900™ II SFM
Gibco™

Sf-900™ II SFM

Sf-900™ II SFM is a serum-free, protein-free insect cell culture medium optimized for the growth and maintenance of Spodoptera frugiperdaRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1090217910 L
1090218720 L
109021615 L
Catalog number 10902179
Price (USD)
1,032.65
Online Exclusive
1,054.00
Save 21.35 (2%)
Each
Quantity:
10 L
Customize this product
Price (USD)
1,032.65
Online Exclusive
1,054.00
Save 21.35 (2%)
Each

Sf-900™ II SFM is a serum-free, protein-free insect cell culture medium optimized for the growth and maintenance of Spodoptera frugiperda (Sf9 and Sf21) cells and for large-scale production of recombinant proteins expressed using the baculovirus expression vector system (BEVS). This medium is suitable for suspension and monolayer culture methods and supports growth of other lepidopteran cell lines. Sf-900™ II SFM features:

  • Superior long-term, high-density growth
  • Optimized for recombinant protein production
  • Serum-free, protein-free, ready-to-use formulation
  • Scalable in bioreactors

Superior long-term, high-density growth

Spodoptera frugiperda (Sf9) cells grown in Sf-900™ II SFM achieve maximum cell densities of 9 to 12 x 106 cells/mL, a significant improvement over competitors' formulations and Grace’s medium (see Product Manual). Increases in maximum cell densities of 20–100% are also observed with the Lymantria dispar (Gypsy moth) and Trichoplusia ni (Tn-368; Cabbage Looper) cell lines. Sf-900™ II SFM is capable of supporting the cultures to >20 passages.

Optimized for recombinant protein production

Traditionally, Grace's medium supplemented with 10% FBS has been used for recombinant protein expression. Sf-900™ II SFM is an improved serum-free, protein-free medium designed for growth of Sf9 and other lepidopteran cell lines and production of insect virus and rDNA proteins.

Serum-free, protein-free, ready-to-use formulation

Sf-900™ II SFM is a serum-free, protein-free medium that allows for much easier purification of your protein of interest. Sf-900™ II SFM is ready to use; it does not require addition of serum, glutamine, or surfactants. Cells adapted to other commercially available serum-free media can be subcultured directly into Sf-900™ II SFM, usually without any further adaptation. Cells usually require some adaptation from serum-containing formulations.

Scalable in bioreactors

The utility of Sf-900™ II SFM in larger-scale cell culture systems was demonstrated with a 5 L Celligen™ bioreactor. Successful infections with rAcNPV were carried out producing rβ-Gal and rEPO (see Product Manual).

Product use

Customers using Gibco™ Sf-900 II SFM in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).

cGMP manufacturing and quality system

For supply chain continuity, we manufacture Sf-900 II SFM at two separate facilities located in Grand Island, NY and Scotland, UK. Both sites are compliant with cGMP manufacturing requirements, are certified to ISO 13485, and are registered with the FDA as medical device manufacturers.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Specifications
Cell LineSf21, Sf9
Cell TypeInsect Cell
Product LineGibco, Sf-900, SynerGy
Product TypeInsect Cell Serum Free Medium (SFM)
Quantity10 L
Shipping ConditionRoom Temperature
SpeciesS. frugiperda, Spodoptera frugiperda
ClassificationProtein-free, Serum-free
FormLiquid
Serum LevelSerum-free
With AdditivesGlutamine
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Frequently asked questions (FAQs)

Can Sf9 cells in Sf-900 III SFM (Cat. No. 12659017) be thawed and grown in Sf-900 II SFM instead?

It should be okay to thaw the cells into Sf-900 II SFM. This is a richer media compared to the Sf-900 III SFM so the cells would have an easy time adapting. We would recommend taking the cells through 3 passages in the new medium before using them for any experiments as that they have enough time to adapt.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What antimicrobials can be used in insect culture and at what concentration?

Many antibiotics are suitable for use with insect cells. The following antibiotics are commonly used:
- Penicillin/Streptomycine: 50-100 U/mL; 50-100 µg/mL
- Amphotericin B (Fungizone antimycotic): 0.25 µg/mL
- Gentamicin: 0.5 mL of 10 mg/mL solution in 500 mL media (final concentration: 10 µg/mL)

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Is it necessary to heat-inactivate my serum before adding it to my medium?

Heat inactivation is not necessary. Our team has routinely used serum that has not been heat-inactivated, and we have not observed any effect on cell growth or morphology.

Many cells do not require heat-inactivated FBS. Some cells prefer heat-inactivated FBS. For instance, we use heat-inactivated FBS for our insect cell lines, i.e., Sf9 and Sf21 cells.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Do you offer serum-free insect media?

Yes, we offer a variety of serum-free insect media. Please go here (http://www.thermofisher.com/us/en/home/life-science/cell-culture/insect-cell-culture/insect-cell-culture-misc/serum-free-media.html?icid=cvc-insect-media-c2t2) to view the media we offer and the differences between them.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Will your company continue to support other films?

Yes. Although we feel that the PE film exceeds many of the technical properties of the other available films, we will continue to offer and support our other currently available films through custom orders.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.