Chlorodiphenylphosphine, 95%, tech.
Chlorodiphenylphosphine, 95%, tech.
Chlorodiphenylphosphine, 95%, tech.
Thermo Scientific Chemicals

Chlorodiphenylphosphine, 95%, tech.

CAS: 1079-66-9 | C12H10ClP | 220.64 g/mol
数量:
25 mL
100 mL
500 mL
パッケージ:
Glass bottle
製品番号(カタログ番号) 109135000
価格(JPY)
-
数量:
500 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1079-66-9
IUPAC Namechlorodiphenylphosphane
Molecular FormulaC12H10ClP
InChI KeyXGRJZXREYAXTGV-UHFFFAOYSA-N
SMILESClP(C1=CC=CC=C1)C1=CC=CC=C1
さらに表示
Appearance (Color)Colorless to yellow
Appearance (Form)Liquid
AppearanceMay develop some turbidity or precipitate
Infrared spectrumConforms
GC>=94 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only