Chemical Identifiers
CAS327-97-9
IUPAC Name(1S,3R,4R,5R)-3-{[(2E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy}-1,4,5-trihydroxycyclohexane-1-carboxylic acid
Molecular FormulaC16H18O9
InChI KeyCWVRJTMFETXNAD-JUHZACGLSA-N
SMILESO[C@@H]1C[C@](O)(C[C@@H](OC(=O)\C=C\C2=CC=C(O)C(O)=C2)[C@@H]1O)C(O)=O
View more