Interested in a bulk or custom version of this product? Request pricing.

TIMP1 Recombinant Human Protein

Catalog number: 10934H08H50

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

TIMP metallopeptidase inhibitor 1 (TIMP1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human TIMP1 (NP_003245.1) (Met 1-Ala 207).

Activity: Measured by its ability to inhibit human MMP2 cleavage of a fluorogenic peptide substrate MCA-PLGL-DPA-AR-NH2. The IC50 value is approximately 6 nM.

Formulation: Lyophilized in 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: TIMP1M, RP1-230G1.3, CLGI, EPA, EPO, FLJ90373, HCI, TIMP
Gene Symbol: TIMP1
Molecular Weight: 22 kDa
Product Line: Sino Biological®
Product Size: 50 µg
Protein Family: Proteases Peptidases & Inhibitors
Protein Name: TIMP meallopeptidase inhibitor 1
Protein Subtype: TIMP
Protein Tag or Fusion: Untagged
Purity: ≥ 90% by SDS-PAGE
Research Category: Angiogenesis, Inflammation, Neurobiology, Oncology
Species: Human

Contents & storage

Includes 1 vial of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols