1-(Chloromethyl)naphthalene, 95%
1-(Chloromethyl)naphthalene, 95%
1-(Chloromethyl)naphthalene, 95%
Thermo Scientific Chemicals

1-(Chloromethyl)naphthalene, 95%

CAS: 86-52-2 | C11H9Cl | 176.65 g/mol
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 109451000
Price (JPY)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS86-52-2
IUPAC Name1-(chloromethyl)naphthalene
Molecular FormulaC11H9Cl
InChI KeyXMWGTKZEDLCVIG-UHFFFAOYSA-N
SMILESClCC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Remainder2-Chloromethylnaphtalene
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only