4-Chloro-3-methylphenol, 99+%
4-Chloro-3-methylphenol, 99+%
4-Chloro-3-methylphenol, 99+%
Thermo Scientific Chemicals

4-Chloro-3-methylphenol, 99+%

CAS: 59-50-7 | C7H7ClO | 142.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 109481000
Price (USD)
34.00
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
34.00
Each
Chemical Identifiers
CAS59-50-7
IUPAC Name4-chloro-3-methylphenol
Molecular FormulaC7H7ClO
InChI KeyCFKMVGJGLGKFKI-UHFFFAOYSA-N
SMILESCC1=CC(O)=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=99.0 %
Appearance (Color)White to almost white
Melting point64°C to 67°C
Appearance (Form)Crystals, flakes, pellets or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only