Interested in a bulk or custom version of this product? Request pricing.

A2M Recombinant Human Protein, His tag

Catalog number: 10952H08B5

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Alpha-2-Macroglobulin (A2M), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymetic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human A2M (NP_000005.2) (Met 1-Ala 1474) fused to a polyhistidine tag at the C-terminus.

Activity: Measured by its ability to trap trypsin. The trapped trypsin is no longer able to interact with protein substrates or inhibitors, but still able to cleave small peptide substrates or inhibitors.The IC50 value is <5 nM.

Formulation: 20 mM Tris, 500 mM NaCl, pH 7.4.

Reconstitution: No reconstitution needed. Store at -80°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Endotoxin Level: < 1 EU/µg
Expression: Insect
Form: Frozen
Gene Aliases: A2mp
Gene Symbol: A2M
Molecular Weight: 164 kDa
Product Line: Sino Biological®
Product Size: 5 µg
Protein Family: Cell Structure⁄Organelle Proteins, Phosphatases & Inhibitors
Protein Name: alpha-2-macroglobulin
Protein Subtype: Protease Inhibitors
Protein Tag or Fusion: His Tag
Purity: ≥ 90% by SDS-PAGE
Research Category: Other Research, Signal Transduction
Species: Human

Contents & storage

Store at -80 °C.

Documents

Manuals & protocols