1-Chloro-2-nitrobenzene, 99+%
1-Chloro-2-nitrobenzene, 99+%
1-Chloro-2-nitrobenzene, 99+%
Thermo Scientific Chemicals

1-Chloro-2-nitrobenzene, 99+%

CAS: 88-73-3 | C6H4ClNO2 | 157.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Catalog number 109620010
Price (USD)
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS88-73-3
IUPAC Name1-chloro-2-nitrobenzene
Molecular FormulaC6H4ClNO2
InChI KeyBFCFYVKQTRLZHA-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Infrared spectrumConforms
Appearance (Form)Fused crystalline mass
Melting point32°C to 34°C
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only