5-chloro-2-nitrobenzoic acid, 99%
5-chloro-2-nitrobenzoic acid, 99%
5-chloro-2-nitrobenzoic acid, 99%
Thermo Scientific Chemicals

5-chloro-2-nitrobenzoic acid, 99%

CAS: 2516-95-2 | C7H4ClNO4 | 201.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
Packaging:
View all
Glass bottle
Catalog number 109680100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2516-95-2
IUPAC Name5-chloro-2-nitrobenzoic acid
Molecular FormulaC7H4ClNO4
InChI KeyZKUYSJHXBFFGPU-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(Cl)=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to yellow-green
Loss on drying=<0.5 %
Appearance (Form)Crystalline powder
HPLC>=98.5 %
Melting point136°C to 139°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only