UltraPure™ DNase/RNase-Free Distilled Water
Invitrogen™

UltraPure™ DNase/RNase-Free Distilled Water

UltraPure™ DNase/RNase-Free Distilled Water is designed for use in all molecular biology applications. It is 0.1-μm membrane-filtered and tested forRead more
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
10 x 500 mL
Catalog number 10977023
Price (MXN)
6,818.98
Each
Quantity:
10 x 500 mL
Request bulk or custom format
Price (MXN)
6,818.98
Each

UltraPure™ DNase/RNase-Free Distilled Water is designed for use in all molecular biology applications. It is 0.1-μm membrane-filtered and tested for DNase and RNase activity.

Performance and quality testing

No DNase, RNase, or protease activity detected. Our distilled water system is routinely monitored for compliance with current USP monograph test requirements for Water for Injection (WFI).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
PackagingBottle
Product LineUltraPure
Purification MethodMembrane-Filtered
Recommended Storage• 10 bottles of UltraPure DNase/RNase-Free Distilled Water (500 mL/bottle)

Store at room temperature.

Treatment(s)Not DEPC-Treated
GradeMolecular Biology
Quantity10 x 500 mL
Unit SizeEach

Frequently asked questions (FAQs)

Does Thermo Fisher Scientific offer a protease-free DNase?

We do not test for protease activity as part of our QC but there is PMSF (a protease inhibitor) in the storage buffer. Furthermore, in the preparation of DNase I, we use a soybean trypsin inhibitor column to remove proteases.

What is the USP conductivity specification for UltraPure DNase/RNase-Free Distilled Water?

We perform daily testing of the water according to USP specifications. The USP specification is that conductivity be “not more than 1 micromho/cm”.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Do you offer DNase- and RNAse-free distilled water?

Yes, we offer UltraPure DNase/RNase-Free Distilled Water (Cat. No. 10977015 or 10977023).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (21)

Citations & References
Abstract
Identification of the sex-determining locus of Atlantic salmon (Salmo salar) on chromosome 2.
Authors:Artieri CG, Mitchell LA, Ng SH, Parisotto SE, Danzmann RG, Hoyheim B, Phillips RB, Morasch M, Koop BF, Davidson WS,
Journal:Cytogenet Genome Res
PubMed ID:16276105
'We have integrated data from linkage mapping, physical mapping and karyotyping to gain a better understanding of the sex-determining locus, SEX, in Atlantic salmon (Salmo salar). SEX has been mapped to Atlantic salmon linkage group 1 (ASL1) and is associated with several microsatellite markers. We have used probes designed from ... More
Oligonucleotide microarray analysis of genomic imbalance in children with mental retardation.
Authors:Friedman JM, Baross A, Delaney AD, Ally A, Arbour L, Armstrong L, Asano J, Bailey DK, Barber S, Birch P, Brown-John M, Cao M, Chan S, Charest DL, Farnoud N, Fernandes N, Flibotte S, Go A, Gibson WT, Holt RA, Jones SJ, Kennedy GC, Krzywinski M, Langlois S, Li HI, McGillivray BC, Nayar T, Pugh TJ, Rajcan-Separovic E, Schein JE, Schnerch A, Siddiqui A, Van Allen MI, Wilson G, Yong SL, Zahir F, Eydoux P, Marra MA,
Journal:Am J Hum Genet
PubMed ID:16909388
'The cause of mental retardation in one-third to one-half of all affected individuals is unknown. Microscopically detectable chromosomal abnormalities are the most frequently recognized cause, but gain or loss of chromosomal segments that are too small to be seen by conventional cytogenetic analysis has been found to be another important ... More
Beringian origins and cryptic speciation events in the fly agaric (Amanita muscaria).
Authors:Geml J, Laursen GA, O'neill K, Nusbaum HC, Taylor DL,
Journal:Mol Ecol
PubMed ID:16367842
'Amanita muscaria sensu lato has a wide geographic distribution, occurring in Europe, Asia, Africa, Australia, New Zealand, and North, Central and South America. Previous phylogenetic work by others indicates three geographic clades (i.e. ''Eurasian'', ''Eurasian-alpine'' and ''North American'' groups) within A. muscaria. However, the historical dispersal patterns of A. muscaria ... More
RNA interference of the clock gene period disrupts circadian rhythms in the cricket Gryllus bimaculatus.
Authors:Moriyama Y, Sakamoto T, Karpova SG, Matsumoto A, Noji S, Tomioka K,
Journal:J Biol Rhythms
PubMed ID:18663238
'Periodic expression of so-called clock genes is an essential part of the circadian clock. In Drosophila melanogaster the cyclic expression of per and tim through an autoregulatory feedback loop is believed to play a central role in circadian rhythm generation. However, it is still elusive whether this hypothesis is applicable ... More
PCR detection of staphylococcal enterotoxin genes in Staphylococcus aureus strains isolated from raw and pasteurized milk.
Authors:Rall VL, Vieira FP, Rall R, Vieitis RL, Fernandes A, Candeias JM, Cardoso KF, Araújo JP,
Journal:Vet Microbiol
PubMed ID:18572331
'Milk is considered a nutritious food because it contains several important nutrients including proteins and vitamins. Conversely, it can be a vehicle for several pathogenic bacteria such as Staphylococcus aureus. This study aimed to analyze the frequency of genes encoding the staphylococcal enterotoxins (SEs) SEA, SEB, SEC, SED, SEE, SEG, ... More