Mitotane
Thermo Scientific Chemicals

Mitotane

CAS: 53-19-0 | C14H10Cl4 | 320.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 109850050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS53-19-0
IUPAC Name1-chloro-4-[2,2-dichloro-1-(2-chlorophenyl)ethyl]benzene
Molecular FormulaC14H10Cl4
InChI KeyJWBOIMRXGHLCPP-UHFFFAOYNA-N
SMILESClC(Cl)C(C1=CC=C(Cl)C=C1)C1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Melting point74°C to 80°C
Infrared spectrumConforms
HPLC>=96.0 %
Loss on drying=<1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only