3-Chloropropionyl chloride, 98%
3-Chloropropionyl chloride, 98%
3-Chloropropionyl chloride, 98%
Thermo Scientific Chemicals

3-Chloropropionyl chloride, 98%

CAS: 625-36-5 | C3H4Cl2O | 126.96 g/mol
数量:
100 mL
500 mL
パッケージ:
Glass bottle
製品番号(カタログ番号) 109995000
価格(JPY)
-
数量:
500 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS625-36-5
IUPAC Name3-chloropropanoyl chloride
Molecular FormulaC3H4Cl2O
InChI KeyINUNLMUAPJVRME-UHFFFAOYSA-N
SMILESClCCC(Cl)=O
さらに表示
GC>=97.5 %
Appearance (Color)Clear colorless to dark brown
Infrared spectrumConforms
Refractive index1.4550 to 1.4590 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only