Thermo Scientific™
Explosion-Proof Refrigerator/Freezers
Related applications:
Cold Storage
Prevent the triggering of an explosion inside or outside of the unit with Thermo Scientific™ Explosion-Proof Refrigerator/Freezers. Suitable for use in Class I, Division I Group C and D hazardous environments, this device meets OSHA and National Fire Protection Association safety requirements.
Back to top
  • Thermostat and compressor components are isolated within a vapor-proof enclosure
  • Electrical connections are hardwired to prevent sparking and accidental disconnection
  • Magnetic vinyl gaskets for positive door seal CFC-free insulation and refrigerant
  • Manual defrost
  • Warranty varies by country, please contact us for details